Assay Details
Target Gene Details
Entrez Gene ID: | 27303 |
Gene Name: | RNA binding motif single stranded interacting protein 3 |
Gene Aliases: |
- |
Location: |
Chr.3:29280860-30010395 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBMS3 | NM_001003792.2 | NP_001003792.1 | ||
| NM_001003793.2 | NP_001003793.1 | |||
| NM_001177711.1 | NP_001171182.1 | |||
| NM_001177712.1 | NP_001171183.1 | |||
| NM_014483.3 | NP_055298.2 | |||
| XM_005265060.2 | XP_005265117.1 | |||
| XM_005265061.2 | XP_005265118.1 | |||
| XM_005265062.1 | XP_005265119.1 | |||
| XM_017006178.1 | XP_016861667.1 | |||
| XM_017006179.1 | XP_016861668.1 | |||
| XM_017006180.1 | XP_016861669.1 | |||
| XM_017006181.1 | XP_016861670.1 | |||
| XM_017006182.1 | XP_016861671.1 | |||
| AF023259.1 | AAC63910.1 | |||
| AK292789.1 | ||||
| AY236871.1 | AAP75555.1 | |||
| AY236872.1 | AAP75556.1 | |||
| BC117315.1 | ||||
| BC143521.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010530 | Chr.3:29245678 - 30074618 on Build GRCh38 | Gain |
|
| nsv590026 | Chr.3:29246160 - 29579113 on Build GRCh38 | Gain |
|
| esv2759137 | Chr.3:29142404 - 29653997 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map