Assay Details
Target Gene Details
Entrez Gene ID: | 132200 |
Gene Name: | chromosome 3 open reading frame 49 |
Gene Aliases: |
- |
Location: |
Chr.3:63819365-63848636 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C3orf49 | NR_026866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008751 | Chr.3:63595517 - 63862172 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map