Assay Details
Target Gene Details
Entrez Gene ID: | 401067 |
Gene Name: | IQ motif containing F3 |
Gene Aliases: |
- |
Location: |
Chr.3:51826883-51830858 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IQCF3 | NM_001085479.2 | NP_001078948.1 | ||
| NM_001207023.1 | NP_001193952.1 | |||
| BC146874.1 | ||||
| BC146889.1 | ||||
| BC171811.1 | ||||
| BC171818.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001350 | Chr.3:51826801 - 51865950 on Build GRCh38 | Gain |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|

Genomic Map