Assay Details
Target Gene Details
Entrez Gene ID: | 80145 |
Gene Name: | THO complex 7 |
Gene Aliases: |
NIF3L1BP1, fSAP24, hTREX30 |
Location: |
Chr.3:63833870-63864478 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THOC7 | NM_001285387.2 | NP_001272316.1 | ||
| NM_001285404.1 | NP_001272333.1 | |||
| NM_025075.3 | NP_079351.2 | |||
| XM_006713339.3 | XP_006713402.1 | |||
| AF104914.1 | ||||
| AI391685.1 | ||||
| AI784549.1 | ||||
| AK027098.1 | BAB15656.1 | |||
| AK225110.1 | ||||
| BC065012.1 | AAH65012.1 | |||
| BI914906.1 | ||||
| CR457373.1 | CAG33654.1 | |||
| HY023143.1 |
More Information
Set Membership: |
|

Genomic Map