Assay Details
Target Gene Details
Entrez Gene ID: | 60678 |
Gene Name: | eukaryotic elongation factor, selenocysteine-tRNA specific |
Gene Aliases: |
EFSEC, SELB |
Location: |
Chr.3:128153456-128426193 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEFSEC | NM_021937.4 | NP_068756.2 | ||
| XM_005247696.2 | XP_005247753.1 | |||
| XM_017006975.1 | XP_016862464.1 | |||
| XM_017006976.1 | XP_016862465.1 | |||
| XM_017006977.1 | XP_016862466.1 | |||
| XM_017006980.1 | XP_016862469.1 | |||
| AF268872.1 | AAG13375.1 | |||
| BC007933.2 | AAH07933.1 | |||
| BG420135.1 | ||||
| DB454660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002685 | Chr.3:127819453 - 128455596 on Build GRCh38 | Gain |
|
| esv32985 | Chr.3:128124111 - 128192840 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140176123] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map