Assay Details
Target Gene Details
Entrez Gene ID: | 387 |
Gene Name: | ras homolog family member A |
Gene Aliases: |
ARH12, ARHA, RHO12, RHOH12 |
Location: |
Chr.3:49359136-49412097 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOA | NM_001313941.1 | NP_001300870.1 | ||
| NM_001313943.1 | NP_001300872.1 | |||
| NM_001313944.1 | NP_001300873.1 | |||
| NM_001313945.1 | NP_001300874.1 | |||
| NM_001313946.1 | NP_001300875.1 | |||
| NM_001313947.1 | NP_001300876.1 | |||
| NM_001664.3 | NP_001655.1 | |||
| AF498970.1 | AAM21117.1 | |||
| AK130066.1 | ||||
| AK130808.1 | ||||
| AK222556.1 | BAD96276.1 | |||
| AK296654.1 | ||||
| AK307220.1 | ||||
| AW403464.1 | ||||
| BC001360.2 | AAH01360.1 | |||
| BC005976.1 | AAH05976.1 | |||
| BF966305.1 | ||||
| BQ721697.1 | ||||
| BT019870.1 | AAV38673.1 | |||
| BU509012.1 | ||||
| BX647063.1 | CAE46190.1 | |||
| L09159.1 | AAA50612.1 | |||
| L25080.1 | AAC33178.1 | |||
| LN607834.1 | ||||
| X05026.1 | CAA28690.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv236940 | Chr.3:49362038 - 49367342 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map