Assay Details
Target Gene Details
Entrez Gene ID: | 4045 |
Gene Name: | limbic system-associated membrane protein |
Gene Aliases: |
IGLON3, LAMP |
Location: |
Chr.3:115802363-116445542 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSAMP | NM_001318915.1 | NP_001305844.1 | ||
| NM_002338.4 | NP_002329.2 | |||
| XM_011512840.2 | XP_011511142.1 | |||
| XM_017006383.1 | XP_016861872.1 | |||
| AK315286.1 | ||||
| BC033803.1 | AAH33803.1 | |||
| BM041903.1 | ||||
| U41901.1 | AAC50569.1 |
Target Gene Details
Entrez Gene ID: | 101926903 |
Gene Name: | LSAMP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:116360024-116370085 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LSAMP-AS1 | NR_109998.1 | |||
| BX110108.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759170 | Chr.3:116366831 - 116403101 on Build GRCh38 | Loss |
|
| nsv437338 | Chr.3:116357442 - 116391788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map