Assay Details
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2-AS1 | NR_110202.1 | |||
| NR_110203.1 | ||||
| DB454604.1 | ||||
| HY022483.1 | ||||
| HY205836.1 |
Target Gene Details
Entrez Gene ID: | 201651 |
Gene Name: | arylacetamide deacetylase pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151770456-151784894 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACP1 | NR_026915.1 | 3 | 350 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522472 | Chr.3:151743597 - 151864407 on Build GRCh38 | Loss |
|
| esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss |
|
| dgv4919n100 | Chr.3:151776103 - 151841876 on Build GRCh38 | Loss |
|

Genomic Map