Assay Details
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AADACL2-AS1 | NR_110202.1 | |||
NR_110203.1 | ||||
DB454604.1 | ||||
HY022483.1 | ||||
HY205836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv522472 | Chr.3:151743597 - 151864407 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
