Assay Details
Target Gene Details
| Entrez Gene ID: | 152330 | 
| Gene Name: | contactin 4 | 
| Gene Aliases: | AXCAM, BIG-2 | 
| Location: | Chr.3:2098771-3059080 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 31 | 
Target Gene Details
| Entrez Gene ID: | 100873975 | 
| Gene Name: | CNTN4 antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.3:3039661-3061145 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CNTN4-AS1 | NR_046554.1 | |||
| BM920658.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv589371 | Chr.3:2170634 - 3594980 on Build GRCh38 | Loss |  LOC100130207  CNTN4  TRNT1  CNTN4-AS1  IL5RA  CRBN | 
| esv2751994 | Chr.3:2200757 - 3178886 on Build GRCh38 | Gain |  CNTN4  TRNT1  CNTN4-AS1  IL5RA  CRBN | 
| esv2422274 | Chr.3:2100396 - 3314907 on Build GRCh38 | Duplication |  LOC100130207  CNTN4  TRNT1  CNTN4-AS1  CNTN4-AS2  IL5RA  CRBN | 
| nsv1011872 | Chr.3:2782874 - 3057776 on Build GRCh38 | Gain |  CNTN4  CNTN4-AS1 | 
| nsv521796 | Chr.3:3038825 - 3151219 on Build GRCh38 | Loss |  CNTN4  TRNT1  CNTN4-AS1  IL5RA  CRBN | 
| dgv4636n100 | Chr.3:2825734 - 3271845 on Build GRCh38 | Gain |  LOC100130207  CNTN4  TRNT1  CNTN4-AS1  IL5RA  CRBN | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top