Assay Details
Target Gene Details
Entrez Gene ID: | 25827 |
Gene Name: | F-box and leucine rich repeat protein 2 |
Gene Aliases: |
FBL2, FBL3 |
Location: |
Chr.3:33277422-33422371 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXL2 | NM_001171713.1 | NP_001165184.1 | ||
| NM_012157.3 | NP_036289.3 | |||
| XM_005265014.2 | XP_005265071.1 | |||
| XM_005265015.3 | XP_005265072.1 | |||
| XM_005265016.2 | XP_005265073.1 | |||
| XM_005265017.3 | XP_005265074.1 | |||
| XM_011533559.2 | XP_011531861.1 | |||
| XM_017006092.1 | XP_016861581.1 | |||
| XM_017006093.1 | XP_016861582.1 | |||
| XM_017006094.1 | XP_016861583.1 | |||
| AF174589.1 | AAF04510.1 | |||
| AF176518.1 | AAF03128.1 | |||
| AF186273.1 | AAD56248.1 | |||
| AK001271.1 | ||||
| AK001438.1 | BAA91691.1 | |||
| AK294209.1 | ||||
| AK297534.1 | ||||
| AK298967.1 | ||||
| AK299665.1 | ||||
| AK302218.1 | ||||
| AK303764.1 | ||||
| AK316229.1 | ||||
| AK316383.1 | ||||
| AU124816.1 | ||||
| BC031556.1 | AAH31556.1 | |||
| CR457121.1 | CAG33402.1 | |||
| DA031508.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759138 | Chr.3:33253615 - 33429981 on Build GRCh38 | Gain |
|
| nsv834653 | Chr.3:33225646 - 33381915 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76522528] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map