Assay Details
Target Gene Details
Entrez Gene ID: | 4026 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
- |
Location: |
Chr.3:188153050-188890671 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPP | XM_017006379.1 | XP_016861868.1 | ||
| AL833171.1 |
Target Gene Details
Entrez Gene ID: | 339929 |
Gene Name: | LPP antisense RNA 2 |
Gene Aliases: |
MYCLo-5, MYCLo-6 |
Location: |
Chr.3:188151206-188154088 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPP-AS2 | NR_036497.1 | 1 | 423 | |
| JX046913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34193 | Chr.3:188129943 - 188550793 on Build GRCh38 | Loss |
|
| esv2751987 | Chr.3:188124933 - 188289510 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map