Assay Details
Target Gene Details
Entrez Gene ID: | 4026 |
Gene Name: | LIM domain containing preferred translocation partner in lipoma |
Gene Aliases: |
- |
Location: |
Chr.3:188153050-188890671 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Target Gene Details
Entrez Gene ID: | 100873917 |
Gene Name: | LPP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:188562237-188568666 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LPP-AS1 | NR_046623.1 | |||
| AW592678.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33400 | Chr.3:188360726 - 188682623 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75311616] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map