Assay Details
Target Gene Details
Entrez Gene ID: | 79804 |
Gene Name: | HAND2 antisense RNA 1 (head to head) |
Gene Aliases: |
DEIN, NBLA00301 |
Location: |
Chr.4:173528600-173591255 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAND2-AS1 | NR_003679.2 | 4 | 706 | |
NR_136192.1 | 4 | 4112 | ||
NR_136193.1 | 4 | 4809 | ||
NR_136194.1 | 4 | 4279 | ||
NR_136195.1 | ||||
NR_136196.1 | ||||
NR_136197.1 | 4 | 4183 | ||
NR_136198.1 | 3 | 4000 | ||
NR_136199.1 | 5 | 4322 | ||
NR_136200.1 | 4 | 4812 | ||
NR_136201.1 | 4 | 4979 | ||
AA156933.1 | 1 | 178 | ||
AK054605.1 | ||||
AK123668.1 | ||||
DA863153.1 | ||||
DB292439.1 | ||||
DQ831955.1 | 1 | 2872 | ||
DQ831956.1 | 1 | 2872 | ||
EF529495.1 | 4 | 3112 | ||
EF529496.1 | 4 | 3035 | ||
EF529497.1 | 4 | 2983 | ||
EF529498.1 | 4 | 3247 | ||
EF529499.1 | 4 | 613 | ||
EF529500.1 | 4 | 503 | ||
EF529501.1 | 3 | 298 | ||
EF529502.1 | 3 | 501 | ||
EF529503.1 | 4 | 780 | ||
EF529504.1 | 5 | 822 | ||
EF529505.1 | 4 | 706 | ||
EU000853.1 | 4 | 4025 | ||
EU000854.1 | 4 | 4025 | ||
EU000855.1 | 3 | 5117 | ||
EU000856.1 | 3 | 5117 |
