Assay Details
Target Gene Details
Entrez Gene ID: | 56884 |
Gene Name: | follistatin like 5 |
Gene Aliases: |
- |
Location: |
Chr.4:161383892-162164034 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSTL5 | NM_001128427.2 | NP_001121899.1 | ||
| NM_001128428.2 | NP_001121900.1 | |||
| NM_020116.4 | NP_064501.2 | |||
| XM_011532126.1 | XP_011530428.1 | |||
| AB033089.1 | ||||
| AK315152.1 | ||||
| BC036502.2 | AAH36502.1 | |||
| DA243182.1 |
Target Gene Details
Entrez Gene ID: | 101928052 |
Gene Name: | uncharacterized LOC101928052 |
Gene Aliases: |
- |
Location: |
Chr.4:162022733-162047550 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101928052 | NR_125888.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015505 | Chr.4:161989825 - 163516137 on Build GRCh38 | Gain |
|
| nsv435789 | Chr.4:162032840 - 162039658 on Build GRCh38 | Deletion |
|
| dgv5454n100 | Chr.4:161000260 - 162613846 on Build GRCh38 | Gain |
|
| nsv1032175 | Chr.4:160266174 - 162269517 on Build GRCh38 | Gain |
|
| nsv595952 | Chr.4:161406415 - 162238893 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map