Assay Details
Target Gene Details
Entrez Gene ID: | 645513 |
Gene Name: | septin 7 pseudogene |
Gene Aliases: |
- |
Location: |
Chr.4:119454783-119499592 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC645513 | NR_037630.1 | |||
BM544557.1 |
