Assay Details
Target Gene Details
Entrez Gene ID: | 493856 |
Gene Name: | CDGSH iron sulfur domain 2 |
Gene Aliases: |
ERIS, Miner1, NAF-1, WFS2, ZCD2 |
Location: |
Chr.4:102868978-102892807 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CISD2 | NM_001008388.4 | NP_001008389.1 | ||
| AK292134.1 | ||||
| BC032300.1 | AAH32300.1 | |||
| BX537971.1 | CAD97935.1 | |||
| DN993861.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759270 | Chr.4:102859548 - 103136164 on Build GRCh38 | Gain |
|
| nsv594957 | Chr.4:102806853 - 102948934 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs72935545] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map