Assay Details
Target Gene Details
Entrez Gene ID: | 401145 |
Gene Name: | coiled-coil serine rich protein 1 |
Gene Aliases: |
FAM190A |
Location: |
Chr.4:90127394-91602219 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830006 | Chr.4:90050505 - 90265189 on Build GRCh38 | Loss |
|
| nsv470053 | Chr.4:89958462 - 90518422 on Build GRCh38 | Gain |
|
| nsv1012406 | Chr.4:89246630 - 90245636 on Build GRCh38 | Loss |
|
| nsv594806 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|

Genomic Map