Assay Details
Target Gene Details
Entrez Gene ID: | 389207 |
Gene Name: | glutaredoxin and cysteine rich domain containing 1 |
Gene Aliases: |
DFNB25, PPP1R88 |
Location: |
Chr.4:42893266-43030658 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GRXCR1 | NM_001080476.2 | NP_001073945.1 | ||
DT932651.1 | ||||
DY654338.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
