Assay Details
Target Gene Details
Entrez Gene ID: | 51306 |
Gene Name: | family with sequence similarity 13 member B |
Gene Aliases: |
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61 |
Location: |
Chr.5:137937909-138051961 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM13B | XM_006714648.1 | XP_006714711.1 | ||
| XM_011543450.1 | XP_011541752.1 | |||
| XM_017009550.1 | XP_016865039.1 |
Target Gene Details
Entrez Gene ID: | 100130172 |
Gene Name: | uncharacterized LOC100130172 |
Gene Aliases: |
- |
Location: |
Chr.5:138032774-138038933 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100130172 | NR_037900.2 | |||
| AK310493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830502 | Chr.5:137909039 - 138106588 on Build GRCh38 | Gain |
|
| esv3606913 | Chr.5:137856857 - 138124533 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map