Assay Details
Target Gene Details
Entrez Gene ID: | 64087 |
Gene Name: | methylcrotonoyl-CoA carboxylase 2 |
Gene Aliases: |
MCCB |
Location: |
Chr.5:71587288-71658706 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCCC2 | NM_022132.4 | NP_071415.1 | ||
| XM_005248567.1 | XP_005248624.1 | |||
| XM_011543529.2 | XP_011541831.1 | |||
| XM_017009688.1 | XP_016865177.1 | |||
| AB050049.1 | BAB16880.1 | |||
| AF261884.1 | AAK49409.1 | |||
| AF301000.1 | AAK16404.1 | |||
| AF310971.1 | AAG53094.1 | |||
| AK310194.1 | ||||
| BC014897.1 | AAH14897.1 | |||
| BC065027.1 | AAH65027.1 | |||
| KU178741.1 | ||||
| KU178742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830340 | Chr.5:71501286 - 71671283 on Build GRCh38 | Loss |
|
| nsv1034703 | Chr.5:71095346 - 71699868 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map