Assay Details
Target Gene Details
Entrez Gene ID: | 101927862 |
Gene Name: | uncharacterized LOC101927862 |
Gene Aliases: |
- |
Location: |
Chr.5:168229583-168232372 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CTB-178M22.2 | NR_109894.1 | |||
N90849.1 |
Target Gene Details
Entrez Gene ID: | 57451 |
Gene Name: | teneurin transmembrane protein 2 |
Gene Aliases: |
ODZ2, TEN-M2, TNM2, ten-2 |
Location: |
Chr.5:166979066-168264159 on Build GRCh38 |
Assay Gene Location: | Within Intron 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TENM2 | NM_001080428.2 | NP_001073897.2 | ||
NM_001122679.1 | NP_001116151.1 | |||
XM_005265950.2 | XP_005266007.1 | |||
XM_005265952.2 | XP_005266009.1 | |||
XM_006714897.2 | XP_006714960.1 | |||
XM_011534604.2 | XP_011532906.1 | |||
XM_017009660.1 | XP_016865149.1 | |||
XM_017009661.1 | XP_016865150.1 | |||
XM_017009662.1 | XP_016865151.1 | |||
XM_017009663.1 | XP_016865152.1 | |||
XM_017009664.1 | XP_016865153.1 | |||
XM_017009665.1 | XP_016865154.1 | |||
XM_017009666.1 | XP_016865155.1 | |||
XM_017009667.1 | XP_016865156.1 | |||
XM_017009668.1 | XP_016865157.1 | |||
XM_017009669.1 | XP_016865158.1 | |||
AB032953.2 | BAA86441.2 | |||
BX648178.1 | ||||
CR936703.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
