Assay Details
Target Gene Details
Entrez Gene ID: | 167227 |
Gene Name: | decapping mRNA 2 |
Gene Aliases: |
NUDT20 |
Location: |
Chr.5:112976702-113022195 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCP2 | NM_001242377.1 | NP_001229306.1 | ||
| NM_152624.5 | NP_689837.2 | |||
| NR_038352.1 | ||||
| XM_017009163.1 | XP_016864652.1 | |||
| AK090564.1 | BAC03479.1 | |||
| AK296361.1 | ||||
| AY135173.1 | AAN08884.1 | |||
| AY146650.1 | AAN62762.1 | |||
| BC064593.1 | AAH64593.1 | |||
| BG720562.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890710 | Chr.5:112940950 - 113923075 on Build GRCh38 | Gain |
|
| nsv599394 | Chr.5:112849794 - 113110626 on Build GRCh38 | Gain |
|
| nsv478144 | Chr.5:112977500 - 112977500 on Build GRCh38 | Novel Sequence Insertion |
|
| nsv1028759 | Chr.5:112964793 - 113022817 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map