Assay Details
Target Gene Details
Entrez Gene ID: | 1719 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
DHFRP1, DYR |
Location: |
Chr.5:80626226-80654981 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHFR | NM_000791.3 | NP_000782.1 | ||
| NM_001290354.1 | NP_001277283.1 | |||
| NM_001290357.1 | NP_001277286.1 | |||
| NR_110936.1 | ||||
| AK293146.1 | ||||
| AK297302.1 | ||||
| AK308492.1 | ||||
| BC000192.1 | AAH00192.1 | |||
| BC003584.1 | AAH03584.2 | |||
| BC070280.1 | AAH70280.1 | |||
| BC071996.1 | AAH71996.1 | |||
| J00140.1 | AAA58485.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483112 | Chr.5:80551019 - 80731514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map