Assay Details
Target Gene Details
Entrez Gene ID: | 168002 |
Gene Name: | dishevelled binding antagonist of beta catenin 2 |
Gene Aliases: |
C6orf116, DAPPER2, DPR2, bA503C24.7 |
Location: |
Chr.6:168292830-168319754 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DACT2 | NM_001286351.1 | NP_001273280.1 | ||
| BC092498.1 | AAH92498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830867 | Chr.6:168203477 - 168373698 on Build GRCh38 | Loss |
|
| esv2733335 | Chr.6:168295536 - 168296633 on Build GRCh38 | Deletion |
|
| esv2759493 | Chr.6:167836188 - 168314704 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map