Assay Details
Target Gene Details
Entrez Gene ID: | 221442 |
Gene Name: | adenylate cyclase 10, soluble pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.6:41101034-41140835 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY10P1 | NR_026938.2 | |||
| AK126092.1 | ||||
| AL833650.1 | ||||
| BX537595.1 | ||||
| BX648193.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5277 | Chr.6:41112016 - 41146182 on Build GRCh38 | Insertion |
|
| nsv602970 | Chr.6:41070039 - 41258521 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149253855] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map