Assay Details
Target Gene Details
Entrez Gene ID: | 346007 |
Gene Name: | eyes shut homolog (Drosophila) |
Gene Aliases: |
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1 |
Location: |
Chr.6:63719980-65707225 on Build GRCh38 |
Assay Gene Location: | Within Intron 45 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYS | NM_001142800.1 | NP_001136272.1 | ||
| NM_001292009.1 | NP_001278938.1 | |||
| FJ416331.1 | ||||
| FM209056.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830674 | Chr.6:63645507 - 63825908 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200327511] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map