Assay Details
Target Gene Details
Entrez Gene ID: | 222663 |
Gene Name: | signal peptide, CUB domain and EGF like domain containing 3 |
Gene Aliases: |
CEGF3 |
Location: |
Chr.6:35214047-35253079 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCUBE3 | NM_001303136.1 | NP_001290065.1 | ||
| NM_152753.3 | NP_689966.2 | |||
| XM_005248943.1 | XP_005249000.1 | |||
| XM_005248947.1 | XP_005249004.1 | |||
| AF452494.1 | AAN76808.1 | |||
| AK291218.1 | ||||
| AY639608.1 | AAU08347.1 | |||
| BC052263.1 | AAH52263.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759419 | Chr.6:35200339 - 35323225 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map