Assay Details
Target Gene Details
Entrez Gene ID: | 29074 |
Gene Name: | mitochondrial ribosomal protein L18 |
Gene Aliases: |
HSPC071, L18mt, MRP-L18 |
Location: |
Chr.6:159789812-159798429 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL18 | NR_134860.1 | |||
| NR_134862.1 | ||||
| NR_134863.1 | ||||
| NR_134864.1 | ||||
| NR_134865.1 | ||||
| NR_134866.1 | ||||
| AK311391.1 | ||||
| AV756740.1 | ||||
| BF797811.1 | ||||
| BU601065.1 | ||||
| CA777760.1 | ||||
| CD675657.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map