Assay Details
Target Gene Details
Entrez Gene ID: | 1235 |
Gene Name: | C-C motif chemokine receptor 6 |
Gene Aliases: |
BN-1, C-C CKR-6, CC-CKR-6, CCR-6, CD196, CKR-L3, CKRL3, CMKBR6, DCR2, DRY6, GPR29, GPRCY4, STRL22 |
Location: |
Chr.6:167111807-167139141 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCR6 | NM_004367.5 | NP_004358.2 | ||
| CD692892.1 | ||||
| DN996219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2752099 | Chr.6:167034693 - 167165614 on Build GRCh38 | Gain |
|
| nsv469673 | Chr.6:167086888 - 167245044 on Build GRCh38 | Loss |
|
| nsv1024657 | Chr.6:167112710 - 167142000 on Build GRCh38 | Loss |
|
| esv2759492 | Chr.6:167087716 - 167776053 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map