Assay Details
Target Gene Details
Entrez Gene ID: | 2941 |
Gene Name: | glutathione S-transferase alpha 4 |
Gene Aliases: |
GSTA4-4, GTA4 |
Location: |
Chr.6:52977948-52995380 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTA4 | NM_001512.3 | NP_001503.1 | ||
XM_005249035.3 | XP_005249092.1 | |||
XM_011514534.2 | XP_011512836.1 | |||
XM_011514535.2 | XP_011512837.1 | |||
AF020918.1 | AAD04711.1 | |||
AF025887.1 | AAC39695.1 | |||
AF125271.1 | AAD27704.1 | |||
AF125272.1 | AAD27705.1 | |||
AF125273.1 | AAD27706.1 | |||
AK054804.1 | ||||
AK315369.1 | ||||
BC015523.2 | AAH15523.1 | |||
BC063439.1 | AAH63439.1 | |||
BG716171.1 | ||||
CR749474.1 | CAH18304.1 | |||
Y13047.1 | CAA73482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759430 | Chr.6:52928056 - 52996406 on Build GRCh38 | Loss |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
