Assay Details
Target Gene Details
Entrez Gene ID: | 154141 |
Gene Name: | membrane bound O-acyltransferase domain containing 1 |
Gene Aliases: |
LPEAT1, LPLAT, LPLAT 1, LPSAT, OACT1, dJ434O11.1 |
Location: |
Chr.6:20099684-20212464 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBOAT1 | NM_001080480.2 | NP_001073949.1 | ||
| NR_073465.1 | ||||
| XM_006714999.2 | XP_006715062.1 | |||
| AB305043.1 | ||||
| AK093994.1 | BAC04264.1 | |||
| AK131269.1 | BAD18447.1 | |||
| AK296857.1 | ||||
| BC150650.1 | ||||
| BC150652.1 |
More Information
Set Membership: |
|

Genomic Map