Assay Details
Target Gene Details
Entrez Gene ID: | 100506409 |
Gene Name: | ELOVL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:11043758-11079144 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELOVL2-AS1 | NR_038962.1 | |||
| NR_038963.1 | ||||
| NR_038964.1 | ||||
| BI828717.1 | ||||
| DN918479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608089 | Chr.6:11059195 - 11087756 on Build GRCh38 | Loss |
|
| nsv1161335 | Chr.6:11071891 - 11097310 on Build GRCh38 | Deletion |
|
| nsv818401 | Chr.6:11073958 - 11083743 on Build GRCh38 | Loss |
|
| esv2670783 | Chr.6:11059326 - 11087887 on Build GRCh38 | Deletion |
|
| nsv441985 | Chr.6:11074069 - 11076965 on Build GRCh38 | Loss |
|
| dgv998n67 | Chr.6:11072086 - 11077368 on Build GRCh38 | Loss |
|
| nsv823424 | Chr.6:11059812 - 11086964 on Build GRCh38 | Loss |
|

Genomic Map