Assay Details
Target Gene Details
Entrez Gene ID: | 245806 |
Gene Name: | vestigial like family member 2 |
Gene Aliases: |
VGL2, VITO1 |
Location: |
Chr.6:117265558-117273565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VGLL2 | NM_153453.1 | NP_703154.1 | ||
| NM_182645.3 | NP_872586.1 | |||
| XM_005266883.2 | XP_005266940.1 | |||
| AJ578053.1 | CAE17331.1 | |||
| AK096878.1 | BAC04881.1 | |||
| AY056583.1 | AAL17720.1 | |||
| BC069316.1 | AAH69316.1 | |||
| BC100798.2 | AAI00799.1 | |||
| BC100799.1 | AAI00800.1 | |||
| BC118545.1 | ||||
| BC118622.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759465 | Chr.6:117110061 - 117306112 on Build GRCh38 | Loss |
|
| nsv830785 | Chr.6:117127780 - 117306112 on Build GRCh38 | Gain |
|
| nsv950498 | Chr.6:117263738 - 117272337 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map