Assay Details
Target Gene Details
Entrez Gene ID: | 594 |
Gene Name: | branched chain keto acid dehydrogenase E1 subunit beta |
Gene Aliases: |
BCKDE1B, BCKDH E1-beta, E1B |
Location: |
Chr.6:80106610-80469088 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCKDHB | NM_000056.4 | NP_000047.1 | ||
| NM_001318975.1 | NP_001305904.1 | |||
| NM_183050.3 | NP_898871.1 | |||
| NR_134945.1 | ||||
| XM_005248756.4 | XP_005248813.1 | |||
| XM_011536023.2 | XP_011534325.1 | |||
| XM_011536024.2 | XP_011534326.1 | |||
| XM_011536025.2 | XP_011534327.1 | |||
| AK289977.1 | ||||
| AK304353.1 | ||||
| AK316545.1 | ||||
| BC040139.2 | AAH40139.1 | |||
| BT020063.1 | AAV38866.1 | |||
| DC372349.1 | ||||
| DC422376.1 | ||||
| M55575.1 | AAA51812.1 | |||
| U50708.1 | AAB16763.1 | |||
| X52446.1 | CAA36685.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5371 | Chr.6:80082685 - 80116423 on Build GRCh38 | Insertion |
|
| esv3890895 | Chr.6:80076658 - 80233186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map