Assay Details
Target Gene Details
Entrez Gene ID: | 57583 |
Gene Name: | transmembrane protein 181 |
Gene Aliases: |
GPR178, KIAA1423 |
Location: |
Chr.6:158536432-158635435 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM181 | NM_020823.1 | NP_065874.1 | ||
| XM_011535999.1 | XP_011534301.1 | |||
| XM_011536000.1 | XP_011534302.1 | |||
| XM_011536003.2 | XP_011534305.1 | |||
| AB037844.1 | ||||
| BC033450.1 | AAH33450.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs114473156] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map