Assay Details
Target Gene Details
Entrez Gene ID: | 6648 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD |
Location: |
Chr.6:159679064-159762529 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOD2 | NM_000636.3 | 5 | 3680 | NP_000627.2 |
| NM_001024465.2 | NP_001019636.1 | |||
| NM_001024466.2 | NP_001019637.1 | |||
| NM_001322814.1 | 4 | 3563 | NP_001309743.1 | |
| NM_001322815.1 | 4 | 3500 | NP_001309744.1 | |
| NM_001322817.1 | NP_001309746.1 | |||
| NM_001322819.1 | 5 | 3586 | NP_001309748.1 | |
| NM_001322820.1 | 5 | 3589 | NP_001309749.1 | |
| AK097395.1 | ||||
| BC012423.1 | AAH12423.1 | |||
| BM724413.1 | ||||
| BM994509.1 | ||||
| BU741675.1 | ||||
| Y00472.1 | CAA68533.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map