Assay Details
Target Gene Details
Entrez Gene ID: | 4698 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A5 |
Gene Aliases: |
B13, CI-13KD-B, CI-13kB, NUFM, UQOR13 |
Location: |
Chr.7:123536997-123601651 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA5 | NM_001282419.2 | NP_001269348.1 | ||
| NM_001282420.2 | NP_001269349.1 | |||
| NM_001282421.2 | NP_001269350.1 | |||
| NM_001282422.2 | NP_001269351.1 | |||
| NM_001291304.1 | NP_001278233.1 | |||
| NM_005000.4 | NP_004991.1 | |||
| NR_104168.2 | ||||
| NR_104169.2 | ||||
| NR_111925.1 | ||||
| NR_111926.1 | ||||
| AK315458.1 | ||||
| AV713284.1 | ||||
| BC000813.1 | AAH00813.1 | |||
| BC020821.1 | AAH20821.1 | |||
| BC070236.1 | AAH70236.1 | |||
| BC070237.1 | AAH70237.1 | |||
| BF671339.1 | ||||
| BF790429.1 | ||||
| BG400546.1 | ||||
| BG611486.1 | ||||
| BT006695.1 | AAP35341.1 | |||
| CR933664.1 | CAI45962.1 | |||
| U53468.1 | AAB02224.1 | |||
| U64028.1 | AAB37259.1 | |||
| W44870.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3614856 | Chr.7:123545800 - 123551524 on Build GRCh38 | Loss |
|
| nsv524192 | Chr.7:122763367 - 123578468 on Build GRCh38 | Gain |
|
| nsv831116 | Chr.7:123447558 - 123619922 on Build GRCh38 | Gain+Loss |
|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map