Assay Details
Target Gene Details
Entrez Gene ID: | 6624 |
Gene Name: | fascin actin-bundling protein 1 |
Gene Aliases: |
FAN1, HSN, SNL, p55 |
Location: |
Chr.7:5592805-5606656 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FSCN1 | NM_003088.3 | NP_003079.1 | ||
| AK095254.1 | ||||
| AK095830.1 | 1 | 213 | ||
| AK316607.1 | ||||
| BC000521.2 | AAH00521.1 | |||
| BC006304.2 | AAH06304.1 | |||
| BC007539.1 | AAH07539.1 | |||
| BC007643.1 | AAH07643.1 | |||
| BC007948.2 | AAH07948.1 | |||
| BC007988.2 | AAH07988.1 | |||
| BT006636.1 | AAP35282.1 | |||
| U03057.1 | AAA86442.1 | |||
| U09873.1 | AAA62201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830890 | Chr.7:5425302 - 5665527 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map