Assay Details
Target Gene Details
Entrez Gene ID: | 100534592 |
Gene Name: | URGCP-MRPS24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:43866558-43906632 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URGCP-MRPS24 | NM_001204871.1 | NP_001191800.1 | ||
| AI027170.1 | ||||
| FY210844.1 | ||||
| FY212401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|

Genomic Map