Assay Details
Target Gene Details
Entrez Gene ID: | 6733 |
Gene Name: | SRSF protein kinase 2 |
Gene Aliases: |
SFRSK2 |
Location: |
Chr.7:105110708-105399366 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRPK2 | NM_001278273.1 | NP_001265202.1 | ||
| NM_182691.2 | NP_872633.1 | |||
| NM_182692.2 | NP_872634.1 | |||
| XM_005250549.3 | XP_005250606.2 | |||
| XM_005250550.3 | XP_005250607.1 | |||
| XM_005250551.4 | XP_005250608.1 | |||
| XM_006716098.2 | XP_006716161.1 | |||
| XM_011516536.2 | XP_011514838.1 | |||
| XM_011516538.2 | XP_011514840.1 | |||
| XM_017012560.1 | XP_016868049.1 | |||
| XM_017012561.1 | XP_016868050.1 | |||
| XM_017012562.1 | XP_016868051.1 | |||
| XM_017012563.1 | XP_016868052.1 | |||
| XM_017012564.1 | XP_016868053.1 | |||
| XM_017012565.1 | XP_016868054.1 | |||
| XM_017012566.1 | XP_016868055.1 | |||
| AK092977.1 | 1 | 1681 | ||
| AK093420.1 | ||||
| AY354201.1 | AAQ63886.1 | |||
| BC035214.1 | AAH35214.1 | |||
| BC068547.1 | AAH68547.1 | |||
| U88666.1 | AAC05299.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv237e55 | Chr.7:105097165 - 105512795 on Build GRCh38 | Gain |
|
| esv2759549 | Chr.7:105117803 - 105568045 on Build GRCh38 | Gain |
|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map