Assay Details
Target Gene Details
Entrez Gene ID: | 7458 |
Gene Name: | eukaryotic translation initiation factor 4H |
Gene Aliases: |
WBSCR1, WSCR1, eIF-4H |
Location: |
Chr.7:74174376-74197101 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF4H | NM_022170.1 | NP_071496.1 | ||
| NM_031992.1 | NP_114381.1 | |||
| XM_011516556.1 | XP_011514858.1 | |||
| XM_011516557.1 | XP_011514859.1 | |||
| AK290676.1 | ||||
| AK297651.1 | ||||
| AK314749.1 | ||||
| BC021214.2 | AAH21214.1 | |||
| D26068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831030 | Chr.7:74111777 - 74250191 on Build GRCh38 | Gain |
|
| esv3613698 | Chr.7:74090462 - 74398730 on Build GRCh38 | Loss |
|
| nsv607480 | Chr.7:74102784 - 74237706 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map