Assay Details
Target Gene Details
Entrez Gene ID: | 6733 |
Gene Name: | SRSF protein kinase 2 |
Gene Aliases: |
SFRSK2 |
Location: |
Chr.7:105110708-105399366 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv237e55 | Chr.7:105097165 - 105512795 on Build GRCh38 | Gain |
|
| esv2759549 | Chr.7:105117803 - 105568045 on Build GRCh38 | Gain |
|
| nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map