Assay Details
Target Gene Details
Entrez Gene ID: | 222234 |
Gene Name: | family with sequence similarity 185 member A |
Gene Aliases: |
- |
Location: |
Chr.7:102748944-102913394 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM185A | XM_006715896.3 | 9 | 1837 | XP_006715959.1 |
| XM_011515923.2 | XP_011514225.1 | |||
| XM_011515924.2 | XP_011514226.1 | |||
| XM_017011846.1 | XP_016867335.1 | |||
| XM_017011847.1 | XP_016867336.1 | |||
| XM_017011848.1 | XP_016867337.1 | |||
| XM_017011849.1 | XP_016867338.1 |
Target Gene Details
Entrez Gene ID: | 222235 |
Gene Name: | F-box and leucine rich repeat protein 13 |
Gene Aliases: |
DRC6, Fbl13 |
Location: |
Chr.7:102699226-103074841 on Build GRCh38 |
Assay Gene Location: | Within Intron 23 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv464667 | Chr.7:102845807 - 102983745 on Build GRCh38 | Loss |
|
| nsv608057 | Chr.7:102799737 - 102898068 on Build GRCh38 | Loss |
|
| esv3614358 | Chr.7:102794927 - 102853299 on Build GRCh38 | Gain |
|
| nsv470382 | Chr.7:102735216 - 102898068 on Build GRCh38 | Loss |
|
| nsv608059 | Chr.7:102830008 - 102864326 on Build GRCh38 | Loss |
|
| dgv6538n100 | Chr.7:102171110 - 103063688 on Build GRCh38 | Gain |
|
| nsv608058 | Chr.7:102826294 - 102931266 on Build GRCh38 | Gain |
|
| esv2752138 | Chr.7:102717873 - 103146688 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75178290] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map