Assay Details
Target Gene Details
Entrez Gene ID: | 6671 |
Gene Name: | Sp4 transcription factor |
Gene Aliases: |
HF1B, SPR-1 |
Location: |
Chr.7:21428071-21514761 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP4 | NM_001326542.1 | NP_001313471.1 | ||
| NM_001326543.1 | NP_001313472.1 | |||
| NM_003112.4 | NP_003103.2 | |||
| XM_005249828.2 | XP_005249885.1 | |||
| XM_005249829.3 | XP_005249886.1 | |||
| XM_011515486.2 | XP_011513788.1 | |||
| XM_017012557.1 | 5 | 2221 | XP_016868046.1 | |
| AK289728.1 | ||||
| BC109300.1 | AAI09301.1 | |||
| BC109301.1 | AAI09302.1 | |||
| X68561.1 | CAA48563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830921 | Chr.7:21449482 - 21648398 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map