Assay Details
Target Gene Details
Entrez Gene ID: | 116988 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
AGAP-3, CENTG3, CRAG, MRIP-1, cnt-g3 |
Location: |
Chr.7:151085831-151144436 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGAP3 | NM_001042535.3 | NP_001036000.1 | ||
| NM_001281300.1 | NP_001268229.1 | |||
| NM_001308304.1 | NP_001295233.1 | |||
| NM_031946.6 | NP_114152.3 | |||
| XM_017011734.1 | XP_016867223.1 | |||
| AB209781.1 | BAD93018.1 | |||
| AF359283.3 | AAK48932.2 | |||
| AF413079.1 | AAL04173.1 | |||
| AK055393.1 | ||||
| DA771207.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv11724n54 | Chr.7:151054503 - 151135826 on Build GRCh38 | Loss |
|
| nsv831188 | Chr.7:151038117 - 151215789 on Build GRCh38 | Loss |
|
| nsv951648 | Chr.7:150931413 - 151106413 on Build GRCh38 | Deletion |
|
| nsv465215 | Chr.7:151065784 - 151128000 on Build GRCh38 | Loss |
|
| dgv6756n100 | Chr.7:151033912 - 151093430 on Build GRCh38 | Gain |
|
| nsv6004 | Chr.7:151067919 - 151098858 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map