Assay Details
Target Gene Details
Entrez Gene ID: | 23333 |
Gene Name: | dpy-19 like 1 (C. elegans) |
Gene Aliases: |
- |
Location: |
Chr.7:34928881-35038041 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPY19L1 | NM_015283.1 | NP_056098.1 | ||
| XM_011515245.2 | XP_011513547.1 | |||
| XM_011515246.2 | XP_011513548.1 | |||
| DQ287932.1 | ABB89208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6320n100 | Chr.7:34875294 - 35092844 on Build GRCh38 | Gain |
|
| nsv830952 | Chr.7:34918149 - 35067312 on Build GRCh38 | Gain |
|
| nsv830953 | Chr.7:34995401 - 35191477 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115006982] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map