Assay Details
Target Gene Details
Entrez Gene ID: | 4697 |
Gene Name: | NDUFA4, mitochondrial complex associated |
Gene Aliases: |
CI-9k, CI-MLRQ, MLRQ |
Location: |
Chr.7:10931953-10940186 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA4 | NM_002489.3 | NP_002480.1 | ||
| AK024837.1 | ||||
| AK130950.1 | ||||
| BC101794.1 | AAI01795.1 | |||
| BC101796.1 | AAI01797.1 | |||
| BC105295.1 | AAI05296.1 | |||
| CR407618.1 | CAG28546.1 | |||
| CR541716.1 | CAG46517.1 | |||
| U94586.1 | AAB52726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161498 | Chr.7:10899112 - 10968568 on Build GRCh38 | Deletion |
|
| esv3891051 | Chr.7:10912986 - 11111729 on Build GRCh38 | Loss |
|
| esv3891050 | Chr.7:10836267 - 11491247 on Build GRCh38 | Gain |
|
| nsv1029882 | Chr.7:10832535 - 11331518 on Build GRCh38 | Loss |
|
| nsv1019888 | Chr.7:10673231 - 11457158 on Build GRCh38 | Gain |
|
| esv2752140 | Chr.7:10777433 - 10990121 on Build GRCh38 | Gain |
|
| nsv606187 | Chr.7:10239859 - 11757275 on Build GRCh38 | Gain |
|
| nsv464364 | Chr.7:10865125 - 10949239 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map