Assay Details
Target Gene Details
Entrez Gene ID: | 155185 |
Gene Name: | archaelysin family metallopeptidase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:2679522-2767486 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMZ1 | NM_133463.3 | NP_597720.1 | ||
| XM_011515151.2 | XP_011513453.1 | |||
| XM_017011775.1 | XP_016867264.1 | |||
| XM_017011776.1 | XP_016867265.1 | |||
| AB075830.1 | ||||
| AK289723.1 | ||||
| BC146753.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605871 | Chr.7:2665632 - 2705490 on Build GRCh38 | Loss |
|
| nsv464257 | Chr.7:2629953 - 2705490 on Build GRCh38 | Loss |
|
| nsv950793 | Chr.7:2592167 - 2703366 on Build GRCh38 | Deletion |
|
| nsv471227 | Chr.7:2589386 - 2729230 on Build GRCh38 | Loss |
|
| dgv11144n54 | Chr.7:2620317 - 2712853 on Build GRCh38 | Loss |
|
| dgv401n21 | Chr.7:2600507 - 2714033 on Build GRCh38 | Loss |
|
| nsv966745 | Chr.7:2671601 - 2682592 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map