Assay Details
Target Gene Details
Entrez Gene ID: | 8379 |
Gene Name: | MAD1 mitotic arrest deficient like 1 |
Gene Aliases: |
MAD1, PIG9, TP53I9, TXBP181 |
Location: |
Chr.7:1815792-2232971 on Build GRCh38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAD1L1 | NM_001013836.1 | NP_001013858.1 | ||
| NM_001013837.1 | NP_001013859.1 | |||
| NM_001304523.1 | NP_001291452.1 | |||
| NM_001304524.1 | NP_001291453.1 | |||
| NM_001304525.1 | NP_001291454.1 | |||
| NM_003550.2 | NP_003541.2 | |||
| XM_005249877.1 | XP_005249934.1 | |||
| XM_011515567.1 | XP_011513869.1 | |||
| XM_011515569.2 | XP_011513871.2 | |||
| XM_011515571.1 | XP_011513873.1 | |||
| XM_017012690.1 | XP_016868179.1 | |||
| XM_017012691.1 | XP_016868180.1 | |||
| AF083811.1 | AAD24498.1 | |||
| AF123318.1 | AAD20359.1 | |||
| AK090959.1 | ||||
| AK314227.1 | ||||
| BC009964.2 | AAH09964.1 | |||
| BU617803.1 | ||||
| U33822.1 | AAC52059.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509186 | Chr.7:1830914 - 1876982 on Build GRCh38 | Insertion |
|
| nsv1030397 | Chr.7:1757834 - 1909030 on Build GRCh38 | Gain |
|
| nsv605833 | Chr.7:1790263 - 2072001 on Build GRCh38 | Loss |
|
| nsv830883 | Chr.7:1797006 - 1969451 on Build GRCh38 | Loss |
|
| nsv1017814 | Chr.7:1772659 - 2237654 on Build GRCh38 | Gain |
|
| nsv950788 | Chr.7:1809865 - 1919564 on Build GRCh38 | Deletion |
|
| nsv471193 | Chr.7:1827280 - 1945404 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map